Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Supplier: Thermo Scientific™ 09.4003
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.Specifications
Pretreatment filter for the Smart2Pure system. Includes filter housing and 1µm filter | |
Smart2Pure, LabTower RO, LabTower TII | |
Pretreatment Filter |