Learn More
PRG2 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00005553T01
Description
Western Blotting
Specifications
293T | |
BC005929 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-PRG2 antibody (H00005553-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Human |
PRG2 293T Cell Transient Overexpression Lysate(Denatured) | |
24.53 | |
pCMV-PRG2 full-length | |
100 uL | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Western Blot |