Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567613
Description
PRH2 Polyclonal Antibody for Western Blot, IHC (P)
The PRH1 gene encodes a member of the heterogeneous family of proline-rich salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature isoforms before secretion from the parotid and submandibular/sublingual glands. In western population this locus is commonly biallelic and encodes proline-rich protein (PRP) isoforms, PRP-1 and PRP-2. The reference genome encodes the PRP-1 allele. Certain alleles of this gene are associated with susceptibility to dental caries. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12.
Specifications
PRH2 | |
Polyclonal | |
Unconjugated | |
PRH2 | |
acidic salivary proline-rich protein, HaeIII type, 2; Db-F; Db-s; PA; Parotid acidic protein; parotid double-band protein; Parotid isoelectric focusing variant protein; parotid proline-rich protein 1/2; Peptide P-C; PIF-F; PIF-S; Pr; Pr1/Pr2; PRH1; PRH2; proline rich protein HaeIII subfamily 1; proline rich protein HaeIII subfamily 2; proline-rich protein HaeIII subfamily 1; proline-rich protein HaeIII subfamily 2; Protein A; Protein C; PRP-1/PRP-2; PRP-3/PRP-4; Salivary acidic proline-rich phosphoprotein 1/2; Salivary acidic proline-rich phosphoprotein 3/4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5555 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P02810 | |
PRH2 | |
Synthesized peptide derived from human PRH2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction