Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Antibody (1-155-2), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179124FR
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Lambda Light Chain | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGLC1 | |
Human IgA1 lambda myeloma protein | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3536 | |
Human | |
Purified |
Flow Cytometry | |
1-155-2 | |
Flow Cytometry | |
IGLC, immunoglobulin lambda constant group | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction