Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD6 Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP17982020UL
Description
CHCHD6 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.Specifications
CHCHD6 | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Affinity Purified | |
CHCHD6 | |
Synthetic peptide directed towards the middle region of human CHCHD6The immunogen for this antibody is CHCHD6. Peptide sequence AAIQDKLFQVAKREREAATKHSKASLPTGEGSISHEEQKSVRLARELESR. | |
26 kDa | |
20ul | |
Store at -20C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Western Blot | |
Western Blot 1:1000 | |
NP_115719 | |
coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016 | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Primary | |
84303 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title