Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 17 Antibody (9F3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242619PCP
Description
Cytokeratin 17 Monoclonal specifically detects Cytokeratin 17 in Human samples. It is validated for Flow Cytometry.Specifications
Cytokeratin 17 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
KRT17 | |
Recombinant human KRT17(1-432aa) purified from E. coli | |
100 μL | |
Cancer | |
3872 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
9F3 | |
Flow Cytometry | |
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction