Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF7 Mouse anti-Human, Alexa Fluor 700, Clone: 3G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242626AF700

Catalog No. NBP242626AI


Add to cart

Description

Description

KLF7 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

KLF7
3G3
Western Blot, ELISA
Purified
KLF7
Recombinant human KLF7 (1-302aa) purified from E. coli
Protein A purified
RUO
Primary
8609
Western Blot, ELISA
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor
Mouse
IgG1 Kappa
100 ul
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit