Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4 Mouse anti-Human, Alexa Fluor 405, Clone: 6D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242642AF405

Catalog No. NBP242642AF


Add to cart

Description

Description

CDK4 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

CDK4
6D10
Western Blot, ELISA
Purified
CDK4
Recombinant human CDK4 (1-303aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Mouse
IgG2a Kappa
100 ul
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers
Primary
1019
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit