Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol Dehydrogenase Mouse anti-Human, DyLight 550, Clone: 10F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242653R

Catalog No. NBP242653R


Add to cart

Description

Description

Sorbitol Dehydrogenase Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Sorbitol Dehydrogenase
10F4
Western Blot, ELISA
Purified
SORD
Recombinant human SORD (1-357aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1
Mouse
IgG1 Kappa
100 ul
Diabetes Research, Lipid and Metabolism
Primary
6652
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit