Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rap2B Mouse anti-Human, HRP, Clone: 62D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242655H

Catalog No. NBP242655H


Add to cart

Description

Description

Rap2B Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Rap2B
62D9
Western Blot, ELISA
Purified
RAP2B
Recombinant human RAP2B (1-183aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
HRP
PBS with No Preservative
MGC20484, RAP2B, member of RAS oncogene family, Ras family small GTP binding protein RAP2B, ras-related protein Rap-2b, small GTP binding protein
Mouse
IgG1 Kappa
100 ul
Signal Transduction
Primary
5912
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit