Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine kinase MT 1A Mouse anti-Human, DyLight 680, Clone: 17A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242664FR

Catalog No. NBP242664FR


Add to cart

Description

Description

Creatine kinase MT 1A Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Creatine kinase MT 1A
17A2
Western Blot, ELISA
Purified
CKMT1A
Recombinant human CKMT1A (40-417aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Acidic-Type Mitochondrial Creatine Kinase, CKMT, CKMT1, CKMT1A, Creatine Kinase U-Type, Mitochondrial, Creatine Kinase, Mitochondrial 1 (Ubiquitous), Creatine Kinase, Mitochondrial 1A, EC 2.7.3.2, Mia-CK, Ubiquitous Mitochondrial Creatine Kinase, U-MtCK
Mouse
IgG2b Kappa
100 ul
Protein Kinase
Primary
548596
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit