Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST13 Mouse anti-Human, DyLight 350, Clone: 5C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242671UV

Catalog No. NBP242671UV


Add to cart

Description

Description

CHST13 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

CHST13
5C6
Western Blot, ELISA
Purified
CHST13
Recombinant human HIP/ST13 (1-369aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
C4ST-3, C4ST3MGC119279, carbohydrate (chondroitin 4) sulfotransferase 13, carbohydrate sulfotransferase 13, Chondroitin 4-O-sulfotransferase 3, Chondroitin 4-sulfotransferase 3, EC 2.8.2.5, MGC119278, MGC119281
Mouse
IgG2a Kappa
100 ul
Lipid and Metabolism
Primary
166012
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit