Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bub3 Mouse anti-Human, Biotin, Clone: 2H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242680B

Catalog No. NBP242680B


Add to cart

Description

Description

Bub3 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Bub3
2H6
Western Blot, ELISA
Purified
BUB3
Recombinant human BUB3 (1-328aa) purified from E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
Biotin
PBS with 0.05% Sodium Azide
BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3
Mouse
IgG2b Kappa
100 ul
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary
9184
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit