Learn More
Pancreatic Polypeptide/PP Mouse anti-Human, DyLight 650, Clone: 6512, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243741C
Catalog No. NBP243741C
Add to cart
Description
Description
Pancreatic Polypeptide/PP Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications
Specifications
Pancreatic Polypeptide/PP
6512
Western Blot
Purified
PPY
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Pancreatic polypeptide. The exact sequence is proprietary.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone
Mouse
IgG2b
0.1 ml
Stem Cell Markers
Primary
5539