Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRIM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548859

Catalog No. PA548859

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DNA primase is the polymerase that synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication.
Specifications

Specifications

PRIM2
Polyclonal
PBS with 0.09% sodium azide
P49643
DNA primase large subunit, 277-, DNA primase 58 kDa subunit, p58, PRIM2, PRIM2A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5558
Western Blot
Unconjugated
PRIM2
Liquid
PRIM2
KLH conjugated synthetic peptide between 317-345 amino acids from the Central region of human PRIM2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit