Learn More
Honeywell North™ Primair™ PA900 Series Accessories Shroud
The Primair™ PA900 Series Accessories Shroud is NIOSH approved and meets your unique needs with the following options: Disposable Long, Disposable Short, Reusable Standard, Reusable Premium.
Manufacturer:  Honeywell North™ PA932AM
 View more versions of this product
Catalog No. 17-000-289
Add to cart
Description
Description
PA900 Series Accessories Shroud, NIOSH approved. Different shroud sizes and materials available for various applications.
  • Disposable Short Shroud is suitable for head and neck protection, chlorine resistant
  • Disposable Long Shroud is suitable for head, neck, and shoulder protection, chlorine resistant
  • Reusable Standard Shroud is suitable for dust and light chemical applications
  • Reusable Premium Shroud is suitable for heavy chemical application
Specifications
Specifications
Reusable Shroud, Standard
PAPR Headgear
Approved for use with the Honeywell North™ PA700 Series PAPR Blower Systems
No
Universal