Learn More
Honeywell North™ Primair™ PA900 Series Accessories Shroud
The Primair™ PA900 Series Accessories Shroud is NIOSH approved and meets your unique needs with the following options: Disposable Long, Disposable Short, Reusable Standard, Reusable Premium.
$37.93 - $100.45
Specifications
Product Type PAPR Headgear
Size Universal
View More Specs
Products 4
Catalog Number Mfr. No. Design Type Disposable Price Quantity & Availability  
Catalog Number Mfr. No. Design Type Disposable Price Quantity & Availability  
17-000-288
View Documents Promotion Details
Honeywell North™
PA9311AM
Disposable Shroud, Short Yes
Each for $37.93
Add to cart
17-000-287
View Documents Promotion Details
Honeywell North™
PA9312AM
Disposable Shroud, Long Yes
Each for $50.23
Add to cart
17-000-289
View Documents Promotion Details
Honeywell North™
PA932AM
Reusable Shroud, Standard No
Each for $74.83
Add to cart
17-000-290
View Documents Promotion Details
Honeywell North™
PA933AM
Reusable Shroud, Premium No
Each for $100.45
Add to cart
Description
Description
PA900 Series Accessories Shroud, NIOSH approved. Different shroud sizes and materials available for various applications.
  • Disposable Short Shroud is suitable for head and neck protection, chlorine resistant
  • Disposable Long Shroud is suitable for head, neck, and shoulder protection, chlorine resistant
  • Reusable Standard Shroud is suitable for dust and light chemical applications
  • Reusable Premium Shroud is suitable for heavy chemical application
Specifications
Specifications
PAPR Headgear
Universal