Learn More
Honeywell North™ Primair™ PA900 Series Hard Hat
The Primair™ PA900 Series Hard Hat ANSI Z89.1-2020, Type 1, Class G features an integrated impact resistant visor, enhanced ergonomics, and more user-friendly headgear. Suitable for use in working areas where eye/face/ear and head protection are required.
$410.00 - $697.00
Specifications
Disposable No
Product Type PAPR Headgear
Size X-Small, Universal
With Exhalation Valve No
View More Specs
Products 5
Catalog Number Mfr. No. Color Design Type Material Includes Price Quantity & Availability  
Catalog Number Mfr. No. Color Design Type Material Includes Price Quantity & Availability  
17-000-281
View Documents
Honeywell North™
PA921
Khaki, Black (Headgear), Clear (Face Shield) Hard Hat with Face Shield Coated Polycarbonate, Anti-scratch, Anti-fog (Visor) Hard Hat; PA032 Breathing Tube; Faceseal
Each for $410.00
Add to cart
17-000-282
View Documents
Honeywell North™
PA921DS
Khaki, Black (Headgear), Clear (Face Shield), White (Shroud) Hard Hat with Face Shield and Disposable Long Shroud Coated Polycarbonate, Anti-scratch, Anti-fog (Visor); Polyester (Shroud) Hard Hat; PA032 Breathing Tube; Disposable, chlorine resistant short shroud suitable for head and neck protection
Each for $522.75
Add to cart
17-000-283
View Documents
Honeywell North™
PA921DL
Black (Headgear), Clear (Face Shield), White (Shroud) Hard Hat with Face Shield and Disposable Short Shroud Coated Polycarbonate, Anti-scratch, Anti-fog (Visor); Polyester Blend (Shroud) Hard Hat; PA032 Breathing Tube; Disposable, chlorine resistant long shroud suitable for head, neck, and shoulder protection
Each for $543.25
Add to cart
17-000-284
View Documents
Honeywell North™
PA922
Khaki, Black (Headgear), Clear (Face Shield) Hard Hat with Face Shield and Premium Shroud Coated Polycarbonate, Anti-scratch, Anti-fog (Visor); Polyester (Shroud) Hard Hat; PA032 Breathing Tube; Reusable standard shroud suitable for dust and light chemical applications
Each for $574.00
Add to cart
17-000-285
View Documents
Honeywell North™
PA923
Black (Headgear), Clear (Face Shield), White (Shroud) Hard Hat with Face Shield and Standard Shroud Coated Polycarbonate, Anti-scratch, Anti-fog (Visor); Flame resistant viscose with silicon coating (Shroud) Hard Hat; PA032 Breathing Tube; Reusable premium shroud suitable for heavy chemical application
Each for $697.00
Add to cart
Description
Description
The Honeywell North™ Primair™ 900 Series is a headgear assembly designed for use with the Honeywell North™ Primair™ 700 Series Belt-Mounted Powered Air Purifying Respirator (PAPR). The PA900 Series headgear solution offers advanced ergonomics, supreme fit and materials, and the highest of protection levels on the market. The PA900 Series is lightweight, easy to don and doff, easy to use/maintain, and adapted with accessories applicable to the task at hand (i.e. shrouds for chemicals).
  • Provides respiratory protection against most dusts, fumes, mists, particulate radio-nuclides, and radon progeny attached to dusts, fumes, and mists, as per configuration selection, for extended periods of time.
  • Ideally suited for applications where non-powered air-purifying respirators are not practical or the higher protection factor of a hood is required, yet where the atmospheres are not immediately dangerous to life and health (IDLH) or where there is an oxygen content of at least 19.5%
  • Meets the requirements of NIOSH CFR 84 when assembled from components listed on the NIOSH approval label included with the product. For detailed information on these systems, see the user manuals for those products. Refer to the NIOSH approval labels provided with the Honeywell North™ Primair™ PA900 Series Headpieces for specific NIOSH approval numbers.
  • The Honeywell North™ Primair™ PA900 Series Headpieces combined with the PA700 Series PAPR is for protection from particulates where (i) the concentration does not exceed the Permissible Exposure Limit (PEL) as determined by the Assigned Protection Factor (APF) of the headpiece, (ii) the concentration is below the contaminant’s IDLH level, and (iii) there is sufficient oxygen in the work environment per the OSHA Regulation 29 CFR Section 1910.134 or CSA Standard Z94.4 requirements.
Specifications
Specifications
No
PAPR Headgear
X-Small, Universal
No