Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning Resources Primary Calculator
SDPSCI_ED

Learning calculator skills is easy.

Manufacturer:  Learning Resources, Inc LER 0037

 View more versions of this product

Catalog No. S79182

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.


Includes: Three-key memory, auto shutoff, eight-digit display, one AA battery (S43921H)

Description

Description

  • Bright blue calculator is both solar and battery powered
  • Students start with basic math operations and work their way up to more advanced concepts such as percents and square roots
  • Perfect for independent math practice
  • Dimensions (W × H): 2.5 × 4.5 in. (6.25 × 11.3cm)

Specifications

Specifications

Primary Calculator
Bright Blue
3-key
3 to 15
11.4cm
6.3cm
Perfect for independent math practice
Digital
2.5 x 4.5 in.
Ea.
4.5 in.
2.5 in.
Battery, Solar
Three-key memory, auto shutoff, eight-digit display, one AA battery (S43921H)
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit