Learn More
Novus Biologicals™ Monkey E-Selectin/CD62E ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192718
Catalog No. NBP192718
Add to cart
Description
Description
The Primate E-Selectin/CD62E ELISA Kit (Colorimetric) measures primate E-Selectin/CD62E in cell culture supernatant, serum (baboon).
Specifications
Specifications
1.9 - 60 U/mL
6401
ELISA Kit
0.4 U/mL
Primate (Pm)
E-Selectin/CD62E
SELE
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sE-selectin, HRP-Conjugate anti-monkey sE-selectin monoclonal antibody, Monkey sE-selectin Standard lyophilized, 120U/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum (baboon)
Storage of components varies. See protocol for specific instructions.
Immunology, Lipid and Metabolism
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell ad