Learn More
Primate IgG, Isotype control, Novus Biologicals™
Monkey Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1970471MG
Catalog No. NBP1970471M
Add to cart
Description
Description
IgG antibody specifically detects IgG in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
IgG
Lyophilized
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
Purified
Monkey
Ion exchange chromatography
RUO
Primary
Monkey
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Reconstitute with 0.5 ml deionized water (or equivalent) to desired concentration.