Learn More
Novus Biologicals™ Human, Monkey IL-12 p70/IL-12A ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192710
Catalog No. NBP192710
Add to cart
Description
Description
The Human, Primate IL-12 p70/IL-12A ELISA Kit (Colorimetric) measures human, primate IL-12 p70/IL-12A in cell culture supernatant, serum (baboon).
Specifications
Specifications
3.9 - 250 U/mL
3592
ELISA Kit
2.6 U/mL
Primate (Pm)
IL-12
IL12A
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-12 p70, Biotin-Conjugate anti-monkey IL-12 p70 monoclonal antibody, Streptavidin-HRP, Monkey IL-12 p70 Standard lyophilized, 500U/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
cell culture supernatant, serum (baboon)
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunology, Innate Immunity
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha cha