Learn More
Novus Biologicals™ Human, Monkey TNF RI/TNFRSF1A ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192722
Catalog No. NBP192722
Add to cart
Description
Description
The Human, Primate TNF RI/TNFRSF1A ELISA Kit (Colorimetric) measures human, primate TNF RI/TNFRSF1A in cell culture supernatant, serum (baboon, cynomolgus).
Specifications
Specifications
0.09 - 6.0 U/mL
7132
ELISA Kit
0.06 U/mL
Primate (Pm)
TNF RI/TNFRSF1A
TNFRSF1A
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sTNF-R I, HRP-Conjugate anti-monkey sTNF-R I monoclonal antibody, Monkey sTNF-R I Standard concentrate, 12U/mL upon dilution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum (baboon, cynomolgus)
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Cytokine Research
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform b