Learn More
Brady™ Prinzing Breaker Lockout
Prevent accidental or unauthorized use of breakers. The Brady™ Prinzing Breaker Lockout is an economic solution that is made of durable polycarbonate material and is quick and easy to apply.
Manufacturer:  Brady™ BL07
 View more versions of this product
Catalog No. 19-103-968
  Check Availability
Add to cart
Specifications
Description
  • All Prinzing circuit breaker lockouts are made of durable polycarbonate material
  • Quick and easy to apply
  • Prevents accidental engagement of circuits
Description & Specifications
Specifications
Clamp-on
4.0 in. x 3.67 in. x 2.5 in.
1/Each
Three phase
Polycarbonate
Red