Learn More
Brady™ Economy Lockout Center, Yellow
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Economy Lockout Center is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ 3003Y
Catalog No. 19-104-023
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Text Legend: LOCKOUT CENTER
14 in.
Acrylic
14 in. x 14 in.
Safety
Black on yellow
(24) heavy duty tag; (4) 1.5 in safety hasp; (6) 0.75 in shackle assorted kd steel padlock
14 in.