Learn More
Brady™ Group Lockout Box
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Group Lockout Box is for group lockout situations involving a large number of workers and equipment. Reduces the number of locks being used and can be used as a portable or mounted lockbox.
Manufacturer:  Brady™ LG096E/DEL
Catalog No. 19-104-096
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Lock box
Text Legend: LOCKOUT BOX THE KEY TO MAKING THE WORKPLACE SAFER
Portable
Safety
Yellow on red
Powder-coated steel