Learn More
Brady™ Lockout Kit, Color-Code Red
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Lockout Kit, Color-Code Red is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ LK628R
Catalog No. 19-104-225
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
(1) lockout box; (1) lockout checklist; (1) osha summary sheet; (1) lockout card; (1) 1.5 in. lockout hasp; (1) kd aluminum padlocks, 1.5 in. shackle, red; (1) lockout keychain; (2) danger sign; (5) danger tag; (1) lockout miniposter
Safety
Red
Powder-coated metal