Learn More
Brady™ Lockout Station
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Lockout Station is a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. Designed with the durability required for an industrial workplace.
Manufacturer:  Brady™ LC139E
Catalog No. 19-104-040
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Black on yellow
(100) cable tie; (2) 2.5 to 5 in gate valve lockout; (2) 3-in-1 plug lockout; (2) no-hole circuit breaker lockout; (2) wall switch lockout; (25) danger tag; (3) multi-pole circuit breaker lockout; (6) 1.5 in lockout hasp; small ball valve lockout
Safety
Tagout station