Learn More
Brady™ Lockout Tagout Station
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Lockout Tagout Station is a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. Designed with the durability required for an industrial workplace.
Manufacturer:  Brady™ LC517E
 View more versions of this product
Catalog No. 19-104-045
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Text Legend: LOCKOUT TAGOUT STATION
30.25 in.
9.5 in.
15 in.
Safety
White
(2) 2.5 to 5 in gate valve lockout; (2) 3-in-1 plug lockout; (25) danger tag; (3) 1.5 in shackle red kd safety padlock; (3) 1.5 in shackle yellow kd safety padlock; (6) 1.5 in safety hasp; small ball valve lockout
Powder-coated steel
9.5 in. x 15 in. x 30.25 in.