Learn More
Brady™ Safety Lockout Center
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Safety Lockout Center is made of tough acrylic plastic and provides ready access to locks, tags, logout devices and information. A successful program begins with the right devices in the right place.
Manufacturer:  Brady™ 3001
Catalog No. 19-104-169
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Mechanical risk lockouts
30 in.
4.5 in.
24 in.
Safety
Black/red on yellow
(1) center board; (2) lockout tags (pk 10 ea); (5) 1.5 in. lockout hasps; (6) kd steel padlocks (1 ea black, blue, green, red, white, yellow); (25) lockout safety booklet; (5) 1.5 in multiple lockout devices
Acrylic
4.5 in. x 24 in. x 30 in.