Learn More
Brady™ Safety Lockout Center, Board Only
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Safety Lockout Center is made of tough acrylic plastic and provides ready access to locks, tags, lockout devices and information.
Supplier: Brady™ 3012
Safety Services Available
Overview
LOTO Assessments
Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Red on White | |
SAFETY LOCK-OUT CENTER USE LOCKS / TAGS TO DISABLE EQUIPMENT PRIOR TO MAINTENANCE, CLEANING OR INSPECTION | |
4.5 in. | |
24 in. |
4.5 x 24 x 30 in. | |
30 in. | |
1 Ea. |