Learn More
Brady™ Small Lockout Kit Pouch
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Small Lockout Kit Pouch is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ LKPOUCH
Catalog No. 19-104-232
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
4 in.
Nylon
7 in. x 4 in.
Safety
Red/white
(1) lockout pouch
7 in.