Brady™ Prinzing Standard MSDS Binders

Used to store company's critical MSDS sheets

Manufacturer: Brady™ 2027

 View more versions of this product

Catalog No. 19-103-215

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Ring 2.5 in.
    Diameter (Metric) Ring 6.3cm
    Includes Polyethylene cover, 91.4cm (36 in.) chain, pair of sheet lifters
    No. of Pages Holds 400 to 500 sheets
    Product Type Bilingual MSDS Binder

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit