Learn More
Brady™ Ten Lock Station, Equipped
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Ten Lock Station is a great way to centralize lockout activity needs by organizing padlocks in one location. Designed with the durability required for an industrial workplace.
Manufacturer:  Brady™ LC205G
Catalog No. 19-104-056
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Text Legend: LOCKOUT STATION
(10) blue kd steel padlock; lockout tag; safety hasp
Safety
Black on yellow