Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prion protein, Mouse anti-Human,Hamster, Clone: 3F4, Novus Biologicals™

Mouse Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Prion protein
Clone 3F4
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Host Species Mouse
Target Species Human, Hamster
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015523
View Documents
Novus Biologicals
NBP2810780.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015524
View Documents
Novus Biologicals
NBP2810780.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Prion protein Monoclonal antibody specifically detects Prion protein in Human, Hamster Samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

Prion protein
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Human, Hamster
Primary
3F4
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit