Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prion protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Prion protein
Applications Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093077
View Documents
Novus Biologicals
NBP303595100UL
100 μg
Each for $320.71
Add to cart
 
NB093078
View Documents
Novus Biologicals
NBP30359520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Prion protein Polyclonal antibody specifically detects Prion protein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, KnockDown
Specifications

Specifications

Prion protein
Unconjugated
Rabbit
Neuroscience, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
5621
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679
Recombinant fusion protein containing a sequence corresponding to amino acids 23-230 of human Prion protein (NP_001073592.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit