Learn More
Invitrogen™ PRKACA (PKA) Recombinant Human Protein
PRKACA (PKA) Recombinant Human Protein
Manufacturer:  Invitrogen™ PR3791B
Encompass
 View more versions of this product
Catalog No. PR3791B
Add to cart
Description
Description
PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.
Specifications
Specifications
NP_002721.1
Dry Ice
Catalytic
Frozen
E. coli
100 µg
PKA Kinase Family
His Tag
Ser⁄Thr Kinases: AGC Group
Kinase Assay
Human
Drug Development
43.5 kDa
PRKACA (PKA)
Yes
Safety and Handling
Safety and Handling
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.