Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKACA (PKA) Recombinant Human Protein Non-distribution product as customer accommodation.

PRKACA (PKA) Recombinant Human Protein

$435.00

Specifications

Accession Number NP_002721.1
Assays PRKACA (PKA) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.4), 24mM MgCl2, 240μM ATP, 180μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name PRKACA (PKA) Recombinant Human Protein
Expression System E. coli
Gene Alias PRKACA (PKA)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
P2912 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
P2912
10μg Each for $435.00
Add to cart
 
Description

Description

PRKACA (PKA) is a serine⁄threonine protein kinase that phosphorylates a broad spectrum of substrates. PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.
Specifications

Specifications

NP_002721.1
PRKACA (PKA) Recombinant Human Protein
PRKACA (PKA)
43.5kDa
1 Each
90% by SDS-PAGE
–80°C
Human
PRKACA (PKA) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.4), 24mM MgCl2, 240μM ATP, 180μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C
E. coli
PKA
PRKACA (PKA) Recombinant Human Protein
PRKACA (PKA) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit