Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCB, Rabbit anti-Human, Clone: A10-F, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15890

Catalog No. 89-998-730


Add to cart

Description

Description

Specifications

Specifications

PRKCB
A10-F
ELISA (1:20000-1:50000) Immunocytochemistry Immunoprecipitation Western Blot (1:5000) The optimal working dilution should be determined by the end user.
protein kinase C, beta
MGC41878/PKC-beta/PKCB/PRKCB1/PRKCB2
Rabbit
IgG
RUO
Antibody
Monoclonal
Human
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide).
Liquid
PRKCB
A synthetic peptide corresponding to amino acids 658-666 at C-terminus of human PRKCB1.
100 μL
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
5579
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit