Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCE Mouse anti-Canine, Human, Clone: OTI1B4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502417S
Description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.Specifications
PRKCE | |
Monoclonal | |
0.88 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q02156 | |
PKCE, nPKC-epsilon | |
Mouse | |
IgG2b | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
5581, 609934 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B4 | |
Unconjugated | |
PRKCE | |
Liquid | |
PRKCE | |
Full length human recombit protein of human PRKCE produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title