Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCQ (PKC theta) Recombinant Human Protein Non-distribution product as customer accommodation.

PRKCQ (PKC theta)

$353.00

Specifications

Accession Number NP_006248
Assays PRKCQ (PKC theta) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 0.1mM EGTA, 1X lipid mix (200μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 100μg/mL PKC epsilon peptide substrate (ERMRPRKRQGSVRRRV) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name PRKCQ (PKC theta) Recombinant Human Protein
Concentration 0.34mg/mL
Expression System Baculovirus
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
P2996 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
P2996
5μg Each for $353.00
Add to cart
 
Description

Description

The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications

Specifications

NP_006248
PRKCQ (PKC theta) Recombinant Human Protein
Baculovirus
PKC
PRKCQ (PKC theta) Recombinant Human Protein
PRKCQ (PKC theta) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag
PRKCQ (PKC theta) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 0.1mM EGTA, 1X lipid mix (200μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 100μg/mL PKC epsilon peptide substrate (ERMRPRKRQGSVRRRV) and trace [32P]-γ-ATP for 10 minutes at 30°C
0.34mg/mL
PRKCQ (PKC theta)
84.4kDa
1 Each
90% by SDS-PAGE
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit