Learn More
Abnova™ PRKD2 (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6552
Catalog No. 89-339-455
Add to cart
Description
Description
Human PRKD2 (NP_057541.2, 1 a.a. - 878 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
protein kinase D2
5 µg
HSPC187, PKD2
25865
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.