Learn More
Protein Kinase D2 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI5G1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525230
Catalog No. PIMA525230
Add to cart
Description
Description
Members of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differs among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca 2+. On the other hand, nPKC delta and epsilon, as well as all of the cPKC members, possess phorbol ester-binding activities and kinase activities.
Specifications
Specifications
Protein Kinase D2
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZL6, Q5XIS9
Polycystin 2; Autosomal domit polycystic kidney disease type II protein; Polycystwin; R48321; PKD2
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25865, 292658, 484427, 716736
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI5G1
Unconjugated
PRKD2
Liquid
PRKD2
Full length human recombit protein of PRKD2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat