Learn More
PRKG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 216461AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3533
Add to Cart
Description
Description
Phosphorylates PPP1R12A.
Specifications
Specifications
PRKG1
Polyclonal
Unconjugated
PRKG1
cGK 1, cGK1, CGKI, cGKI alpha, cGKI BETA, PGK, PKG, PRKG1, PRKG1B, PRKGR1A, PRKGR1B, Protein Kinase G1 alpha
Rabbit
Antigen Affinity Chromatography
RUO
19091, 54286, 5592
-20°C
Liquid
Immunoprecipitation, Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0C605, Q13976
Prkg1
PRKG1 Fusion Protein Ag16333
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG