Learn More
PRKG1 Recombinant Human Protein
Manufacturer:  LIFE TECHNOLOGIES PV4340
Encompass
 View more versions of this product
Catalog No. PV4340
Add to cart
Description
Description
PRKG1 is a cGMP-dependent serine⁄threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation
Specifications
Specifications
Kinase Assay
Full Length, Recombinant
Signal Transduction
Human
Dry Ice
PRKG1
Ser⁄Thr Kinases: AGC Group
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
105.3 kDa
PRKG1, PGK
Yes
Liquid
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Kinases & Inhibitors
Baculovirus
Serine⁄Threonine Kinases
protein kinase, cGMP-dependent, type I
NP_006249
5592
10 µg
PKG Kinase Family
GST Tag