Learn More
PRL-2/PTP4A2 Mouse anti-Human, DyLight 350, Clone: 5B6F4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237315UV
Catalog No. NBP237315UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PRL-2/PTP4A2
5B6F4
Western Blot, ELISA
Purified
PTP4A2
Purified recombinant fragment of human PTP4A2 (AA: 58-162) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1.3.48, HU-PP-1HH13, OV-1PTP4A, PRL-2HH7-2, PRL2protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA 2, protein tyrosine phosphatase type IVA, member 2, Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, PTP(CAAXII), PTPCAAX2phosphatase of regenerating liver 2, ptp-IV1a, ptp-IV1b
Mouse
IgG2b
0.1 ml
Primary
8073