Learn More
PRL Mouse anti-Human, Clone: OTI11B11, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA500720
Catalog No. 50-167-1861
Add to cart
Description
Description
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
Specifications
Specifications
PRL
Monoclonal
Unconjugated
PRL
Liquid
PRL
Full length human recombit protein of human PRL produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI11B11
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01236
GHA1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5617