Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRLR Rabbit anti-Human, Unconjugated, Clone: PRLR/3785R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00610

Catalog No. 89-339-589


Add to cart

Description

Description

PRLR Monoclonal antibody specifically detects PRLR in Human samples. It is validated for IHC-P.
Specifications

Specifications

PRLR
PRLR/3785R
prolactin receptor
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
PRLR
hPRLrI
Recombinant protein corresponding to full-length human PRLR
100 μg
Primary
5618
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit