Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT1 Mouse anti-Human, Mouse, FITC, Clone: 779, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243744F

Catalog No. NBP243744F


Add to cart

Description

Description

PRMT1 Monoclonal antibody specifically detects PRMT1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRMT1
779
Immunocytochemistry/Immunofluorescence
Purified
PRMT1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PRMT1. The exact sequence is proprietary.
Protein G purified
Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary
3276
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit