Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT5 Mouse anti-Human, Rat, DyLight 350, Clone: 3610, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243678UV

Catalog No. NBP243678UV


Add to cart

Description

Description

PRMT5 Monoclonal antibody specifically detects PRMT5 in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

PRMT5
3610
Western Blot
Purified
PRMT5
Recombinant protein encompassing a sequence within the center region of human PRMT5. The exact sequence is proprietary.
Protein G purified
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
Primary
10419
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit